Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PAK3 SIRNA Assays ›
          Gene Symbol:
          PAK3
          Gene Name:
          p21 (RAC1) activated kinase 3
          Gene Aliases:
          ARA, MRX30, MRX47, OPHN3, PAK-3, PAK3beta, bPAK, beta-PAK
          Chromosome Location:
          Chr.X: 110944241 - 111227361 on Build GRCh38
          UniGene ID:
          Hs.593599
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n319420

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PAK3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128166.2 NP_001121638.1 16 6340
          NM_001128167.2 NP_001121639.1 15 6267
          NM_001128168.2 NP_001121640.1 20 6589
          NM_001128172.2 NP_001121644.1 15 5942
          NM_001128173.2 NP_001121645.1 19 6526
          NM_001324325.1 NP_001311254.1 16 6418
          NM_001324326.1 NP_001311255.1 16 6263
          NM_001324327.1 NP_001311256.1 18 6375
          NM_001324328.1 NP_001311257.1 17 6308
          NM_001324329.1 NP_001311258.1 19 6523
          NM_001324330.1 NP_001311259.1 16 6415
          NM_001324331.1 NP_001311260.1 17 6330
          NM_001324332.1 NP_001311261.1 18 6481
          NM_001324333.1 NP_001311262.1 18 6456
          NM_001324334.1 NP_001311263.1 17 6411
          NM_002578.4 NP_002569.1 18 6478
          NR_136747.1 18 6505
          NR_136748.1 16 6140
          XM_017029557.1 XP_016885046.1 17 6377
          XM_017029561.1 XP_016885050.1 18 6480
          XM_017029563.1 XP_016885052.1 20 6581
          GenBank mRNA AK124702.1 - 1 2056
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA287714.1 - 1 103 Chr. X - 111224562 - 111225115 [+] on Build GRCh38
          GenBankmRNA DA500564.1 - 1 50 Chr. X - 111224615 - 111225187 [+] on Build GRCh38
          GenBankmRNA DA774189.1 - 1 91 Chr. X - 111224574 - 111225163 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          mitotic cell cycle
          stimulatory C-type lectin receptor signaling pathway
          axonogenesis
          dendrite development
          regulation of actin filament polymerization
          T cell costimulation
          positive regulation of neuron apoptotic process
          ephrin receptor signaling pathway
          synapse organization
          T cell receptor signaling pathway
          dendritic spine morphogenesis
          positive regulation of dendritic spine morphogenesis
          cellular response to organic cyclic compound
          positive regulation of DNA biosynthetic process
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          MAP kinase kinase activity
          protein binding
          ATP binding
          Rho GTPase binding
          SH3 domain binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0