Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADAM22 SIRNA Assays ›
          Gene Symbol:
          ADAM22
          Gene Name:
          ADAM metallopeptidase domain 22
          Gene Aliases:
          ADAM 22, MDC2
          Chromosome Location:
          Chr.7: 87934143 - 88202889 on Build GRCh38
          UniGene ID:
          Hs.256398
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n322242

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADAM22
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001324417.1 NP_001311346.1 30 8709
          NM_001324418.1 NP_001311347.1 32 8994
          NM_001324419.1 NP_001311348.1 32 8991
          NM_001324420.1 NP_001311349.1 31 8907
          NM_001324421.1 NP_001311350.1 29 8588
          NM_016351.5 NP_057435.2 30 8712
          NM_021722.5 NP_068368.2 31 8799
          NM_021723.4 NP_068369.1 31 8820
          XM_005250445.3 XP_005250502.1 30 8668
          XM_006716028.3 XP_006716091.1 32 8866
          XM_006716029.3 XP_006716092.1 29 8544
          XM_011516318.1 XP_011514620.1 33 8997
          XM_011516319.2 XP_011514621.1 33 9081
          XM_011516320.2 XP_011514622.1 31 8779
          XM_011516321.2 XP_011514623.1 31 8755
          XM_011516322.1 XP_011514624.1 33 8910
          XM_011516323.1 XP_011514625.1 32 8802
          XM_011516324.1 XP_011514626.1 33 8669
          XM_017012329.1 XP_016867818.1 33 8853
          XM_017012330.1 XP_016867819.1 32 8850
          XM_017012331.1 XP_016867820.1 32 8883
          XM_017012332.1 XP_016867821.1 32 8799
          XM_017012333.1 XP_016867822.1 30 8588
          XM_017012334.1 XP_016867823.1 32 8766
          XM_017012335.1 XP_016867824.1 32 8742
          XM_017012336.1 XP_016867825.1 31 8715
          XM_017012337.1 XP_016867826.1 30 8531
          XM_017012338.1 XP_016867827.1 31 8791
          XM_017012339.1 XP_016867828.1 31 8655
          XM_017012340.1 XP_016867829.1 22 7728
          XM_017012341.1 XP_016867830.1 29 8332
          GenBank mRNA AL133090.1 - 1 2473
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AI702979.1 - 1 37 Chr. 7 - 88202231 - 88202716 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          cell adhesion
          negative regulation of cell adhesion
          central nervous system development
          adult locomotory behavior
          myelination in peripheral nervous system
          metalloendopeptidase activity
          integrin binding
          protein binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline