Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GCLM SIRNA Assays ›
          Gene Symbol:
          GCLM
          Gene Name:
          glutamate-cysteine ligase modifier subunit
          Gene Aliases:
          GLCLR
          Chromosome Location:
          Chr.1: 93885199 - 93909598 on Build GRCh38
          UniGene ID:
          Hs.315562
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n328477

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GCLM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001308253.1 NP_001295182.1 6 4506
          NM_002061.3 NP_002052.1 7 4572
          XM_011541261.2 XP_011539563.1 7 4373
          XM_017001056.1 XP_016856545.1 8 4774
          XM_017001057.1 XP_016856546.1 7 4708
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BF244123.1 - 1 499 Chr. 1 - 93885613 - 93886176 [-] on Build GRCh38
          GenBankmRNA D62994.1 - 1 178 Chr. 1 - 93885512 - 93885855 [-] on Build GRCh38
          GenBankmRNA D62993.1 - 1 178 Chr. 1 - 93885535 - 93885855 [-] on Build GRCh38
          GenBankmRNA BM821516.1 - 1 459 Chr. 1 - 93885662 - 93886136 [-] on Build GRCh38
          GenBankmRNA BX437592.2 - 1 705 Chr. 1 - 93885583 - 93886379 [-] on Build GRCh38
          GenBankmRNA BM833295.1 - 1 489 Chr. 1 - 93885477 - 93886166 [-] on Build GRCh38
          GenBankmRNA BG503670.1 - 1 448 Chr. 1 - 93885431 - 93886125 [-] on Build GRCh38
          GenBankmRNA CR598364.1 - 1 702 Chr. 1 - 93885236 - 93886379 [-] on Build GRCh38
          GenBankmRNA BM822465.1 - 1 487 Chr. 1 - 93885657 - 93886165 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          sulfur amino acid metabolic process
          cysteine metabolic process
          glutamate metabolic process
          glutathione biosynthetic process
          response to oxidative stress
          apoptotic mitochondrial changes
          positive regulation of glutamate-cysteine ligase activity
          response to drug
          negative regulation of neuron apoptotic process
          regulation of blood vessel size
          response to nitrosative stress
          regulation of mitochondrial depolarization
          negative regulation of extrinsic apoptotic signaling pathway
          glutamate-cysteine ligase activity
          enzyme regulator activity
          glutamate-cysteine ligase catalytic subunit binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0