Search
Search

KCNB1| Gene Symbol: KCNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004975.3 | NP_004966.1 | 2 | 9224 | ||
| XM_006723784.3 | XP_006723847.1 | 3 | 9318 | |||
| XM_011528799.2 | XP_011527101.1 | 3 | 9347 | |||
| GenBank mRNA | AK126976.1 | - | 1 | 1261 | ||