Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUNX1T1 SIRNA Assays ›
Gene Symbol:
RUNX1T1
Gene Name:
RUNX1 translocation partner 1
Gene Aliases:
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location:
Chr.8: 91954967 - 92103365 on Build GRCh38
UniGene ID:
Hs.368431
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n331394
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUNX1T1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198625.1
NP_001185554.1
15
6168
NM_001198626.1
NP_001185555.1
13
6026
NM_001198627.1
NP_001185556.1
14
6086
NM_001198628.1
NP_001185557.1
12
5683
NM_001198629.1
NP_001185558.1
12
5809
NM_001198630.1
NP_001185559.1
13
5869
NM_001198631.1
NP_001185560.1
12
5882
NM_001198632.1
NP_001185561.1
12
5800
NM_001198633.1
NP_001185562.1
11
5771
NM_001198634.1
NP_001185563.1
11
5709
NM_001198679.1
NP_001185608.1
12
5853
NM_004349.3
NP_004340.1
11
5819
NM_175634.2
NP_783552.1
12
5944
NM_175635.2
NP_783553.1
11
5636
NM_175636.2
NP_783554.1
10
5718
XM_006716676.3
XP_006716739.1
13
5753
XM_011517351.2
XP_011515653.1
13
5798
XM_011517352.2
XP_011515654.1
13
6038
XM_011517353.2
XP_011515655.1
12
5746
XM_017013930.1
XP_016869419.1
12
6054
XM_017013931.1
XP_016869420.1
11
5576
XM_017013932.1
XP_016869421.1
13
5941
XM_017013933.1
XP_016869422.1
14
6005
XM_017013934.1
XP_016869423.1
14
6113
XM_017013935.1
XP_016869424.1
13
5918
XM_017013936.1
XP_016869425.1
12
6012
XM_017013937.1
XP_016869426.1
13
5838
XM_017013939.1
XP_016869428.1
7
5469
XM_017013940.1
XP_016869429.1
8
6698
XM_017013941.1
XP_016869430.1
7
6488
GenBank mRNA
AF131817.1
-
1
1379
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
BM931258.1
-
1
470
Chr. 8 - 91956510 - 91957004 [-] on Build GRCh38
GenBankmRNA
Z44064.1
-
154
Chr. - - [-] on Build
GenBankmRNA
DA525131.1
-
1
305
Chr. 8 - 91956313 - 91956872 [-] on Build GRCh38
GenBankmRNA
BX431624.2
-
1
457
Chr. 8 - 91956501 - 91957023 [-] on Build GRCh38
GenBankmRNA
CK301025.1
-
1
419
Chr. 8 - 91956512 - 91956984 [-] on Build GRCh38
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AK162711, BM941168.
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
transcription, DNA-templated
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0