Search
Search

LOC101928068| Gene Symbol: LOC101928068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111907.1 | 2 | 372 | |||
| GenBank mRNA | AK095945.1 | - | 2 | 1397 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | EL946683.1 | - | 1 | 269 | Chr. 1 - 244969448 - 244969900 [-] on Build GRCh38 | |