Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AVP SIRNA Assays ›
Gene Symbol:
AVP
Gene Name:
arginine vasopressin
Gene Aliases:
ADH, ARVP, AVP-NPII, AVRP, VP
Chromosome Location:
Chr.20: 3082555 - 3093521 on Build GRCh38
UniGene ID:
Hs.89648
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n333524
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AVP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_011529267.1
XP_011527569.1
2
1062
GenBank mRNA
AF031475.1
-
1
220
Back To Top
More Information
Panther Classification:
Molecular Function -
neuropeptide
Gene Ontology Categories:
Function(s)
Process(es)
maternal aggressive behavior
positive regulation of systemic arterial blood pressure
renal water homeostasis
generation of precursor metabolites and energy
water transport
apoptotic process
signal transduction
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
negative regulation of female receptivity
grooming behavior
locomotory behavior
positive regulation of cell proliferation
positive regulation of gene expression
positive regulation of glutamate secretion
positive regulation of cell growth
positive regulation of cAMP biosynthetic process
positive regulation of prostaglandin biosynthetic process
positive regulation of cellular pH reduction
positive regulation of peptidyl-serine phosphorylation
response to testosterone
response to nicotine
social behavior
regulation of renal sodium excretion
vasoconstriction
hyperosmotic salinity response
maternal behavior
negative regulation of apoptotic process
penile erection
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
sodium-independent organic anion transport
response to ethanol
positive regulation of vasoconstriction
negative regulation of transmission of nerve impulse
ERK1 and ERK2 cascade
protein kinase C signaling
negative regulation of release of cytochrome c from mitochondria
protein kinase activity
signal transducer activity
receptor binding
neuropeptide hormone activity
neurohypophyseal hormone activity
V1A vasopressin receptor binding
V1B vasopressin receptor binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0