Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AVP SIRNA Assays ›
          Gene Symbol:
          AVP
          Gene Name:
          arginine vasopressin
          Gene Aliases:
          ADH, ARVP, AVP-NPII, AVRP, VP
          Chromosome Location:
          Chr.20: 3082555 - 3093521 on Build GRCh38
          UniGene ID:
          Hs.89648
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n333525

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AVP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011529267.1 XP_011527569.1 2 1221
          GenBank mRNA AF031475.1 - 1 379

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          maternal aggressive behavior
          positive regulation of systemic arterial blood pressure
          renal water homeostasis
          generation of precursor metabolites and energy
          water transport
          apoptotic process
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          negative regulation of female receptivity
          grooming behavior
          locomotory behavior
          positive regulation of cell proliferation
          positive regulation of gene expression
          positive regulation of glutamate secretion
          positive regulation of cell growth
          positive regulation of cAMP biosynthetic process
          positive regulation of prostaglandin biosynthetic process
          positive regulation of cellular pH reduction
          positive regulation of peptidyl-serine phosphorylation
          response to testosterone
          response to nicotine
          social behavior
          regulation of renal sodium excretion
          vasoconstriction
          hyperosmotic salinity response
          maternal behavior
          negative regulation of apoptotic process
          penile erection
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          sodium-independent organic anion transport
          response to ethanol
          positive regulation of vasoconstriction
          negative regulation of transmission of nerve impulse
          ERK1 and ERK2 cascade
          protein kinase C signaling
          negative regulation of release of cytochrome c from mitochondria
          protein kinase activity
          signal transducer activity
          receptor binding
          neuropeptide hormone activity
          neurohypophyseal hormone activity
          V1A vasopressin receptor binding
          V1B vasopressin receptor binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0