Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IKBKB SIRNA Assays ›
          Gene Symbol:
          IKBKB
          Gene Name:
          inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta
          Gene Aliases:
          IKK-beta, IKK2, IKKB, IMD15, NFKBIKB
          Chromosome Location:
          Chr.8: 42271302 - 42332653 on Build GRCh38
          UniGene ID:
          Hs.597664
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n336107

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IKBKB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005273490.2 XP_005273547.1 21 2755
          XM_005273491.4 XP_005273548.1 20 2655
          XM_005273493.3 XP_005273550.1 18 2451
          XM_005273495.2 XP_005273552.1 20 2496
          XM_005273496.3 XP_005273553.1 19 2516
          XM_005273498.3 XP_005273555.1 17 2325
          XM_011544517.2 XP_011542819.1 21 2753
          XM_011544518.2 XP_011542820.1 20 2605
          XM_011544519.2 XP_011542821.1 20 2623
          XM_011544520.2 XP_011542822.1 18 2470
          XM_011544521.2 XP_011542823.1 19 2583
          XM_011544522.2 XP_011542824.1 18 2431
          XM_017013396.1 XP_016868885.1 20 2708
          GenBank mRNA AK095687.1 - 1 241
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA588319.1 - 1 74 Chr. 8 - 42329505 - 42330078 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          stimulatory C-type lectin receptor signaling pathway
          skeletal muscle contraction
          protein phosphorylation
          inflammatory response
          I-kappaB kinase/NF-kappaB signaling
          I-kappaB phosphorylation
          positive regulation of cell proliferation
          response to virus
          response to toxic substance
          regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of neuron projection development
          cortical actin cytoskeleton organization
          neuron projection development
          response to lipopolysaccharide
          tumor necrosis factor-mediated signaling pathway
          negative regulation of myosin-light-chain-phosphatase activity
          TRIF-dependent toll-like receptor signaling pathway
          Fc-epsilon receptor signaling pathway
          regulation of phosphorylation
          response to drug
          serine phosphorylation of STAT protein
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          stress-activated MAPK cascade
          nucleotide-binding oligomerization domain containing signaling pathway
          interleukin-1-mediated signaling pathway
          response to fatty acid
          cellular response to tumor necrosis factor
          establishment of protein localization to plasma membrane
          positive regulation of neuron death
          regulation of establishment of endothelial barrier
          negative regulation of bicellular tight junction assembly
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          IkappaB kinase activity
          protein kinase binding
          protein homodimerization activity
          protein heterodimerization activity
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0