Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKCE SIRNA Assays ›
Gene Symbol:
PRKCE
Gene Name:
protein kinase C epsilon
Gene Aliases:
PKCE, nPKC-epsilon
Chromosome Location:
Chr.2: 45651315 - 46187990 on Build GRCh38
UniGene ID:
Hs.580351
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n336543
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKCE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_011532981.2
XP_011531283.1
1
380
GenBank mRNA
AK022272.1
-
1
156
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
macrophage activation involved in immune response
protein phosphorylation
apoptotic process
cell cycle
cell adhesion
signal transduction
activation of phospholipase C activity
positive regulation of epithelial cell migration
positive regulation of fibroblast migration
positive regulation of cell-substrate adhesion
peptidyl-serine phosphorylation
platelet activation
positive regulation of actin filament polymerization
negative regulation of protein ubiquitination
lipopolysaccharide-mediated signaling pathway
positive regulation of insulin secretion
positive regulation of synaptic transmission, GABAergic
positive regulation of cytokinesis
intracellular signal transduction
locomotory exploration behavior
TRAM-dependent toll-like receptor 4 signaling pathway
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to morphine
positive regulation of MAPK cascade
regulation of peptidyl-tyrosine phosphorylation
positive regulation of lipid catabolic process
release of sequestered calcium ion into cytosol
regulation of release of sequestered calcium ion into cytosol
cell division
regulation of insulin secretion involved in cellular response to glucose stimulus
positive regulation of mucus secretion
cellular response to ethanol
cellular response to prostaglandin E stimulus
cellular response to hypoxia
positive regulation of wound healing
positive regulation of receptor activity
negative regulation of sodium ion transmembrane transporter activity
positive regulation of cellular glucuronidation
actin monomer binding
protein serine/threonine kinase activity
protein kinase C activity
calcium-independent protein kinase C activity
signal transducer activity
protein binding
ATP binding
enzyme activator activity
enzyme binding
receptor activator activity
ethanol binding
metal ion binding
14-3-3 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0