Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAP2K6 SIRNA Assays ›
          Gene Symbol:
          MAP2K6
          Gene Name:
          mitogen-activated protein kinase kinase 6
          Gene Aliases:
          MAPKK6, MEK6, MKK6, PRKMK6, SAPKK-3, SAPKK3
          Chromosome Location:
          Chr.17: 69414697 - 69553854 on Build GRCh38
          UniGene ID:
          Hs.463978
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n342561

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAP2K6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005257515.2 XP_005257572.1 12 12124
          XM_005257516.2 XP_005257573.1 12 11997
          XM_006721975.3 XP_006722038.1 12 12025
          XM_011525025.2 XP_011523327.1 12 12411
          XM_011525026.2 XP_011523328.1 12 11872
          XM_011525027.2 XP_011523329.1 12 11787
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI918782.1 - 1 501 Chr. 17 - 69552044 - 69552846 [+] on Build GRCh38
          GenBankmRNA BC037328.1 - 1 1163 Chr. 17 - 69551358 - 69553861 [+] on Build GRCh38
          GenBankmRNA BM811540.1 - 1 285 Chr. 17 - 69552244 - 69553130 [+] on Build GRCh38
          GenBankmRNA DA893572.1 - 1 151 Chr. 17 - 69552370 - 69552891 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          response to ischemia
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          DNA damage induced protein phosphorylation
          cell cycle arrest
          signal transduction
          peptidyl-tyrosine phosphorylation
          ovulation cycle process
          positive regulation of prostaglandin secretion
          proteolysis in other organism
          response to drug
          positive regulation of apoptotic process
          positive regulation of nitric-oxide synthase biosynthetic process
          cardiac muscle contraction
          nucleotide-binding oligomerization domain containing signaling pathway
          cellular response to sorbitol
          receptor signaling protein serine/threonine kinase activity
          MAP kinase kinase activity
          protein tyrosine kinase activity
          protein binding
          ATP binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0