Search
Search

KCND3| Gene Symbol: KCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005270851.4 | XP_005270908.1 | 8 | 6737 | ||
| XM_006710629.3 | XP_006710692.1 | 8 | 7047 | |||
| XM_006710630.3 | XP_006710693.1 | 7 | 6680 | |||
| XM_017001244.1 | XP_016856733.1 | 8 | 7062 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF070632.1 | - | 2 | 618 | Chr. 1 - 111770662 - 111772569 [-] on Build GRCh38 | |