Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIP1 SIRNA Assays ›
          Gene Symbol:
          HIP1
          Gene Name:
          huntingtin interacting protein 1
          Gene Aliases:
          HIP-I, ILWEQ, SHON, SHONbeta, SHONgamma
          Chromosome Location:
          Chr.7: 75533298 - 75738976 on Build GRCh38
          UniGene ID:
          Hs.329266
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n351999

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HIP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243198.2 NP_001230127.1 29 7149
          NM_005338.6 NP_005329.3 31 7302
          XM_005250304.2 XP_005250361.1 31 7303
          XM_005250305.2 XP_005250362.1 31 7308
          XM_011516116.2 XP_011514418.1 32 6889
          XM_017012099.1 XP_016867588.1 31 7223
          GenBank mRNA BC082970.1 - 1 378
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY796763.2 - 1 320 Chr. 7 - 75533759 - 75534355 [-] on Build GRCh38
          GenBankmRNA CX788913.1 - 1 255 Chr. 7 - 75533488 - 75534283 [-] on Build GRCh38
          GenBankmRNA BE621970.1 - 1 288 Chr. 7 - 75533586 - 75534324 [-] on Build GRCh38
          GenBankmRNA BU849461.1 - 1 378 Chr. 7 - 75533644 - 75534413 [-] on Build GRCh38
          GenBankmRNA CB999243.1 - 1 343 Chr. 7 - 75533701 - 75534377 [-] on Build GRCh38
          GenBankmRNA BX115118.1 - 1 116 Chr. 7 - 75533428 - 75534151 [-] on Build GRCh38
          GenBankmRNA CX784523.1 - 1 544 Chr. 7 - 75533800 - 75534579 [-] on Build GRCh38
          GenBankmRNA CN278847.1 - 1 253 Chr. 7 - 75533610 - 75534283 [-] on Build GRCh38
          GenBankmRNA CN278846.1 - 1 273 Chr. 7 - 75533515 - 75534303 [-] on Build GRCh38
          GenBankmRNA AA479868.1 - 1 116 Chr. 7 - 75533715 - 75534145 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cytoskeleton organization
          cell differentiation
          regulation of apoptotic process
          positive regulation of receptor-mediated endocytosis
          clathrin coat assembly
          clathrin-mediated endocytosis
          apoptotic signaling pathway
          actin binding
          structural constituent of cytoskeleton
          protein binding
          clathrin binding
          phosphatidylinositol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0