Search
Search

SCN1A| Gene Symbol: SCN1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011511602.2 | XP_011509904.1 | 1 | 21 | ||
| XM_011511605.2 | XP_011509907.1 | 1 | 29 | |||
| XM_011511606.2 | XP_011509908.1 | 1 | 21 | |||
| XM_017004644.1 | XP_016860133.1 | 1 | 21 | |||
| XM_017004645.1 | XP_016860134.1 | 1 | 29 | |||
| XM_017004646.1 | XP_016860135.1 | 1 | 35 | |||
| XM_017004647.1 | XP_016860136.1 | 1 | 28 | |||
| XM_017004650.1 | XP_016860139.1 | 1 | 23 | |||
| XM_017004652.1 | XP_016860141.1 | 1 | 16 | |||
| XM_017004653.1 | XP_016860142.1 | 1 | 34 | |||
| XM_017004654.1 | XP_016860143.1 | 1 | 45 | |||
| GenBank mRNA | DQ993523.1 | - | 1 | 40 | ||