Search
Search

SCN1A| Gene Symbol: SCN1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202435.1 | NP_001189364.1 | 1 | 27 | ||
| XM_017004648.1 | XP_016860137.1 | 1 | 57 | |||
| XM_017004649.1 | XP_016860138.1 | 1 | 111 | |||
| XM_017004651.1 | XP_016860140.1 | 1 | 49 | |||
| GenBank mRNA | DQ993522.1 | - | 1 | 172 | ||
| EU368117.1 | - | 1 | 27 | |||