Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARRB1 SIRNA Assays ›
          Gene Symbol:
          ARRB1
          Gene Name:
          arrestin beta 1
          Gene Aliases:
          ARB1, ARR1
          Chromosome Location:
          Chr.11: 75260122 - 75351831 on Build GRCh38
          UniGene ID:
          Hs.503284
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n361295

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARRB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004041.4 NP_004032.2 16 5222
          NM_020251.3 NP_064647.1 15 5198
          XM_011545034.2 XP_011543336.1 17 5106
          XM_011545035.2 XP_011543337.1 16 5082
          XM_017017750.1 XP_016873239.1 18 5178
          XM_017017751.1 XP_016873240.1 17 5154
          XM_017017752.1 XP_016873241.1 17 5134
          XM_017017753.1 XP_016873242.1 17 5198
          XM_017017754.1 XP_016873243.1 16 5110
          GenBank mRNA AL157484.1 - 1 45
          DA358665.1 - 1 224

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          positive regulation of protein phosphorylation
          G-protein coupled receptor internalization
          positive regulation of receptor internalization
          transcription from RNA polymerase II promoter
          apoptotic process
          phototransduction
          protein transport
          protein ubiquitination
          platelet activation
          negative regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of protein binding
          negative regulation of interleukin-6 production
          negative regulation of interleukin-8 production
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of GTPase activity
          positive regulation of smooth muscle cell apoptotic process
          positive regulation of Rho protein signal transduction
          positive regulation of histone acetylation
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          response to antidepressant
          follicle-stimulating hormone signaling pathway
          stress fiber assembly
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of GTPase activity
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of histone H4 acetylation
          histone acetyltransferase activity
          enzyme inhibitor activity
          GTPase activator activity
          insulin-like growth factor receptor binding
          protein binding
          transcription factor binding
          estrogen receptor binding
          mitogen-activated protein kinase kinase binding
          ubiquitin protein ligase binding
          alpha-1A adrenergic receptor binding
          alpha-1B adrenergic receptor binding
          angiotensin receptor binding
          follicle-stimulating hormone receptor binding
          V2 vasopressin receptor binding
          AP-2 adaptor complex binding
          clathrin adaptor activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          transcription regulatory region DNA binding
          ion channel binding
          protein phosphorylated amino acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0