Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCNI SIRNA Assays ›
          Gene Symbol:
          CCNI
          Gene Name:
          cyclin I
          Gene Aliases:
          CCNI1, CYC1, CYI
          Chromosome Location:
          Chr.4: 77047155 - 77076013 on Build GRCh38
          UniGene ID:
          Hs.518827
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n363376

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCNI
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011531555.1 XP_011529857.1 5 2001
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CD677085.1 - 1 149 Chr. 4 - 77047244 - 77047869 [-] on Build GRCh38
          GenBankmRNA BM474423.1 - 1 536 Chr. 4 - 77047352 - 77048259 [-] on Build GRCh38
          GenBankmRNA BQ441592.1 - 1 572 Chr. 4 - 77047506 - 77048292 [-] on Build GRCh38
          GenBankmRNA BU177849.1 - 1 376 Chr. 4 - 77047286 - 77048087 [-] on Build GRCh38
          GenBankmRNA AV734353.1 - 1 287 Chr. 4 - 77047333 - 77048006 [-] on Build GRCh38
          GenBankmRNA BU184352.1 - 1 471 Chr. 4 - 77047379 - 77048191 [-] on Build GRCh38
          GenBankmRNA CD110649.1 - 1 476 Chr. 4 - 77047460 - 77048196 [-] on Build GRCh38
          GenBankmRNA D54275.1 - 1 298 Chr. 4 - 77047673 - 77048017 [-] on Build GRCh38
          GenBankmRNA BP261800.1 - 1 563 Chr. 4 - 77047702 - 77048283 [-] on Build GRCh38
          GenBankmRNA AJ713357.1 - 1 143 Chr. 4 - 77047675 - 77047863 [-] on Build GRCh38
          GenBankmRNA BU187633.1 - 1 386 Chr. 4 - 77047232 - 77048106 [-] on Build GRCh38
          GenBankmRNA CN403766.1 - 1 159 Chr. 4 - 77047252 - 77047879 [-] on Build GRCh38
          GenBankmRNA BU932363.1 - 1 477 Chr. 4 - 77047469 - 77048196 [-] on Build GRCh38
          GenBankmRNA CF455619.1 - 1 475 Chr. 4 - 77047448 - 77048169 [-] on Build GRCh38
          GenBankmRNA CN415135.1 - 1 414 Chr. 4 - 77047529 - 77048134 [-] on Build GRCh38
          GenBankmRNA CN403764.1 - 1 374 Chr. 4 - 77047664 - 77048092 [-] on Build GRCh38
          GenBankmRNA BP262085.1 - 1 563 Chr. 4 - 77047703 - 77048283 [-] on Build GRCh38
          GenBankmRNA BU571052.1 - 1 478 Chr. 4 - 77047403 - 77048198 [-] on Build GRCh38
          GenBankmRNA AW951424.1 - 1 263 Chr. 4 - 77047421 - 77047983 [-] on Build GRCh38
          GenBankmRNA AW021333.1 - 1 245 Chr. 4 - 77047617 - 77047960 [-] on Build GRCh38
          GenBankmRNA CB115271.1 - 1 327 Chr. 4 - 77047514 - 77048047 [-] on Build GRCh38
          GenBankmRNA BP423340.1 - 1 285 Chr. 4 - 77047512 - 77048004 [-] on Build GRCh38
          GenBankmRNA BP417704.1 - 1 367 Chr. 4 - 77047628 - 77048087 [-] on Build GRCh38
          GenBankmRNA DA599007.1 - 1 403 Chr. 4 - 77047559 - 77048123 [-] on Build GRCh38
          GenBankmRNA BG896208.1 - 1 335 Chr. 4 - 77047510 - 77048020 [-] on Build GRCh38
          GenBankmRNA BU194859.1 - 1 285 Chr. 4 - 77047257 - 77048005 [-] on Build GRCh38
          GenBankmRNA BM687996.1 - 1 147 Chr. 4 - 77047294 - 77047859 [-] on Build GRCh38
          GenBankmRNA BQ220811.1 - 1 568 Chr. 4 - 77047436 - 77048288 [-] on Build GRCh38
          GenBankmRNA BQ638675.1 - 1 221 Chr. 4 - 77047377 - 77047932 [-] on Build GRCh38
          GenBankmRNA BE892565.1 - 1 263 Chr. 4 - 77047416 - 77047983 [-] on Build GRCh38
          GenBankmRNA BM685707.1 - 1 142 Chr. 4 - 77047293 - 77047855 [-] on Build GRCh38
          GenBankmRNA BQ435048.1 - 1 285 Chr. 4 - 77047177 - 77048005 [-] on Build GRCh38
          GenBankmRNA C16167.1 - 1 137 Chr. 1 - 149486993 - 149487164 [+] on Build GRCh38
          GenBankmRNA CD639310.1 - 1 358 Chr. 4 - 77047414 - 77048076 [-] on Build GRCh38
          GenBankmRNA BF982889.1 - 1 150 Chr. 4 - 77047162 - 77047870 [-] on Build GRCh38
          GenBankmRNA BM827353.1 - 1 182 Chr. 4 - 77047346 - 77047902 [-] on Build GRCh38
          GenBankmRNA H02742.1 - 1 124 Chr. 4 - 77047460 - 77047844 [-] on Build GRCh38
          GenBankmRNA CD299221.1 - 1 334 Chr. 4 - 77047349 - 77048051 [-] on Build GRCh38
          GenBankmRNA BU178811.1 - 1 399 Chr. 4 - 77047260 - 77048119 [-] on Build GRCh38
          GenBankmRNA BM703279.1 - 1 320 Chr. 4 - 77047580 - 77048039 [-] on Build GRCh38
          GenBankmRNA BP432211.1 - 1 196 Chr. 4 - 77047461 - 77047916 [-] on Build GRCh38
          GenBankmRNA CD356153.1 - 1 474 Chr. 4 - 77047466 - 77048196 [-] on Build GRCh38
          GenBankmRNA BG165160.1 - 1 271 Chr. 4 - 77047360 - 77047992 [-] on Build GRCh38
          GenBankmRNA AV734949.1 - 1 338 Chr. 4 - 77047370 - 77048057 [-] on Build GRCh38
          GenBankmRNA AA725244.1 - 1 280 Chr. 4 - 77047649 - 77047991 [-] on Build GRCh38
          GenBankmRNA BQ186504.1 - 1 339 Chr. 4 - 77047565 - 77048058 [-] on Build GRCh38
          GenBankmRNA AL550901.3 - 1 212 Chr. 4 - 77047155 - 77047932 [-] on Build GRCh38
          GenBankmRNA CR615035.1 - 1 212 Chr. 4 - 77047155 - 77047932 [-] on Build GRCh38
          GenBankmRNA CD520883.1 - 1 312 Chr. 4 - 77047353 - 77048030 [-] on Build GRCh38
          GenBankmRNA CN415136.1 - 1 296 Chr. 4 - 77047287 - 77048016 [-] on Build GRCh38
          GenBankmRNA CB998394.1 - 1 397 Chr. 4 - 77047441 - 77048116 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s)

          spermatogenesis
          regulation of cell cycle

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-k5tsx:80/100.66.78.246:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0