Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022145.1 | - | 2069 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY073511.1 | - | 1 | 85 | Chr. 17 - 63585572 - 63586065 [+] on Build GRCh38 |