Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MOB1A SIRNA Assays ›
          Gene Symbol:
          MOB1A
          Gene Name:
          MOB kinase activator 1A
          Gene Aliases:
          C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B
          Chromosome Location:
          Chr.2: 74152598 - 74178967 on Build GRCh38
          UniGene ID:
          Hs.602092
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n367242

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MOB1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001317110.1 NP_001304039.1 6 3282
          NM_001317111.1 NP_001304040.1 6 3178
          NM_001317112.1 NP_001304041.1 5 3121
          NM_018221.4 NP_060691.2 6 3285
          XM_011532948.2 XP_011531250.1 5 2981
          GenBank mRNA AL122062.1 - 1 1000
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA046075.1 - 1 367 Chr. 2 - 74154151 - 74154594 [-] on Build GRCh38
          GenBankmRNA DA074754.1 - 1 190 Chr. 2 - 74153837 - 74154415 [-] on Build GRCh38
          GenBankmRNA BQ228447.1 - 1 293 Chr. 2 - 74153747 - 74154518 [-] on Build GRCh38
          GenBankmRNA BQ921235.1 - 1 374 Chr. 2 - 74153684 - 74154600 [-] on Build GRCh38
          GenBankmRNA AU140106.1 - 1 122 Chr. 2 - 74153691 - 74154348 [-] on Build GRCh38
          GenBankmRNA BM739760.1 - 1 122 Chr. 2 - 74153835 - 74154347 [-] on Build GRCh38
          GenBankmRNA BU191927.1 - 1 368 Chr. 2 - 74153791 - 74154573 [-] on Build GRCh38
          GenBankmRNA BQ930347.1 - 1 254 Chr. 2 - 74153591 - 74154479 [-] on Build GRCh38
          GenBankmRNA BM759194.1 - 1 121 Chr. 2 - 74153998 - 74154347 [-] on Build GRCh38
          GenBankmRNA BU535062.1 - 98 Chr. - - [-] on Build
          GenBankmRNA DB130025.1 - 1 210 Chr. 2 - 74153865 - 74154436 [-] on Build GRCh38
          GenBankmRNA BG118286.1 - 1 366 Chr. 2 - 74153826 - 74154592 [-] on Build GRCh38
          GenBankmRNA BM760831.1 - 1 121 Chr. 2 - 74153997 - 74154347 [-] on Build GRCh38
          GenBankmRNA DA752944.1 - 1 164 Chr. 2 - 74153830 - 74154389 [-] on Build GRCh38
          GenBankmRNA CB267168.1 - 1 351 Chr. 2 - 74154205 - 74154576 [-] on Build GRCh38
          GenBankmRNA BM747834.1 - 1 121 Chr. 2 - 74153747 - 74154347 [-] on Build GRCh38
          GenBankmRNA DA820072.1 - 1 450 Chr. 2 - 74154121 - 74154675 [-] on Build GRCh38
          GenBankmRNA BM791793.1 - 1 544 Chr. 2 - 74154155 - 74154766 [-] on Build GRCh38
          GenBankmRNA DA959704.1 - 1 367 Chr. 2 - 74154020 - 74154593 [-] on Build GRCh38
          GenBankmRNA BE882231.1 - 332 Chr. - - [-] on Build
          GenBankmRNA DA731069.1 - 1 389 Chr. 2 - 74153795 - 74154615 [-] on Build GRCh38
          GenBankmRNA AV714739.1 - 1 144 Chr. 2 - 74153716 - 74154369 [-] on Build GRCh38
          GenBankmRNA BG539117.1 - 403 Chr. - - [-] on Build
          GenBankmRNA CB267169.1 - 1 348 Chr. 2 - 74154209 - 74154576 [-] on Build GRCh38
          GenBankmRNA BQ049002.1 - 1 610 Chr. 2 - 74153998 - 74154838 [-] on Build GRCh38
          GenBankmRNA DA891431.1 - 1 358 Chr. 2 - 74154016 - 74154585 [-] on Build GRCh38
          GenBankmRNA DB107830.1 - 1 388 Chr. 2 - 74154040 - 74154615 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s) Process(es)

          hippo signaling
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0