Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMEM164 SIRNA Assays ›
          Gene Symbol:
          Tmem164
          Gene Name:
          transmembrane protein 164
          Gene Aliases:
          AI316850, AI852450, AW547186, F730011B02
          Chromosome Location:
          Chr.X: 142680347 - 142843494 on Build GRCm38
          UniGene ID:
          Mm.249648
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n408907

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tmem164
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK159942.1 - 1 3606
          AK161476.1 - 1 2809
          BC059043.1 - 1 2905
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BF464704.1 - 1 122 Chr. X - 142838202 - 142838367 [+] on Build GRCm38
          GenBankmRNA BY415605.1 - 1 272 Chr. X - 142838052 - 142838369 [+] on Build GRCm38
          GenBankmRNA BI694207.1 - 1 489 Chr. X - 142837832 - 142838371 [+] on Build GRCm38
          GenBankmRNA BB758691.1 - 1 481 Chr. X - 142837851 - 142838367 [+] on Build GRCm38
          GenBankmRNA BI154518.1 - 1 436 Chr. X - 142837888 - 142838359 [+] on Build GRCm38
          GenBankmRNA CX223200.1 - 135 Chr. - - [-] on Build
          GenBankmRNA BY367312.1 - 1 240 Chr. X - 142838085 - 142838374 [+] on Build GRCm38
          GenBankmRNA BB819399.1 - 1 370 Chr. X - 142837962 - 142838369 [+] on Build GRCm38
          GenBankmRNA CJ309988.1 - 1 350 Chr. X - 142837974 - 142838369 [+] on Build GRCm38
          GenBankmRNA CJ302779.1 - 1 376 Chr. X - 142837948 - 142838369 [+] on Build GRCm38
          GenBankmRNA BY667237.1 - 1 330 Chr. X - 142837994 - 142838369 [+] on Build GRCm38
          GenBankmRNA CF538220.1 - 1 128 Chr. X - 142838196 - 142838371 [+] on Build GRCm38
          GenBankmRNA BY427739.1 - 1 365 Chr. X - 142837960 - 142838369 [+] on Build GRCm38
          GenBankmRNA BY636428.1 - 1 343 Chr. X - 142837982 - 142838369 [+] on Build GRCm38
          GenBankmRNA BB798123.1 - 1 357 Chr. X - 142837967 - 142838358 [+] on Build GRCm38
          GenBankmRNA CN458912.1 - 1 666 Chr. X - 142837657 - 142838351 [+] on Build GRCm38
          GenBankmRNA BY663567.1 - 1 339 Chr. X - 142837986 - 142838369 [+] on Build GRCm38
          GenBankmRNA BY430980.1 - 1 370 Chr. X - 142837954 - 142838369 [+] on Build GRCm38
          GenBankmRNA BB791451.1 - 1 258 Chr. X - 142838092 - 142838367 [+] on Build GRCm38
          GenBankmRNA CJ312834.1 - 1 536 Chr. X - 142837788 - 142838369 [+] on Build GRCm38
          GenBankmRNA CJ314123.1 - 1 343 Chr. X - 142837981 - 142838369 [+] on Build GRCm38
          GenBankmRNA BY484358.1 - 1 317 Chr. X - 142838006 - 142838368 [+] on Build GRCm38
          GenBankmRNA BB820749.1 - 1 373 Chr. X - 142837954 - 142838374 [+] on Build GRCm38
          GenBankmRNA BY477517.1 - 1 475 Chr. X - 142837848 - 142838369 [+] on Build GRCm38
          GenBankmRNA BB801074.1 - 1 340 Chr. X - 142837985 - 142838359 [+] on Build GRCm38
          GenBankmRNA BB811787.1 - 1 376 Chr. X - 142837948 - 142838369 [+] on Build GRCm38
          GenBankmRNA BB804873.1 - 1 346 Chr. X - 142837983 - 142838358 [+] on Build GRCm38
          GenBankmRNA CJ323214.1 - 1 426 Chr. X - 142837898 - 142838369 [+] on Build GRCm38
          GenBankmRNA BY518121.1 - 1 264 Chr. X - 142838115 - 142838367 [+] on Build GRCm38
          GenBankmRNA CJ300662.1 - 1 414 Chr. X - 142837910 - 142838369 [+] on Build GRCm38
          GenBankmRNA CJ235530.1 - 1 648 Chr. X - 142837676 - 142838367 [+] on Build GRCm38

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: BC079345.

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0