Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUP62 SIRNA Assays ›
          Gene Symbol:
          Nup62
          Gene Name:
          nucleoporin 62
          Gene Aliases:
          AA589433, AI426861, AI790512, AU045898, D7Ertd649e, Nupc1, p62
          Chromosome Location:
          Chr.7: 44816420 - 44830807 on Build GRCm38
          UniGene ID:
          Mm.2565
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n424644

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nup62
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK017823.1 - 1353
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BB437859.1 - 1 256 Chr. 7 - 44831547 - 44831834 [+] on Build GRCm38
          GenBankmRNA CJ205005.1 - 1 407 Chr. 7 - 44831396 - 44831833 [+] on Build GRCm38
          GenBankmRNA BB097340.1 - 1 276 Chr. 7 - 44831539 - 44831834 [+] on Build GRCm38
          GenBankmRNA BB143239.1 - 1 270 Chr. 7 - 44831551 - 44831834 [+] on Build GRCm38
          GenBankmRNA BY576017.1 - 1 390 Chr. 7 - 44831417 - 44831834 [+] on Build GRCm38
          GenBankmRNA BB295575.1 - 1 257 Chr. 7 - 44831551 - 44831833 [+] on Build GRCm38
          GenBankmRNA BB141856.1 - 1 237 Chr. 7 - 44831574 - 44831834 [+] on Build GRCm38
          GenBankmRNA BB440375.1 - 1 258 Chr. 7 - 44831545 - 44831834 [+] on Build GRCm38
          GenBankmRNA BY579004.1 - 1 372 Chr. 7 - 44831430 - 44831832 [+] on Build GRCm38
          GenBankmRNA BB145590.1 - 1 287 Chr. 7 - 44831528 - 44831834 [+] on Build GRCm38
          GenBankmRNA BB438338.1 - 1 251 Chr. 7 - 44831558 - 44831834 [+] on Build GRCm38
          GenBankmRNA BB378935.1 - 1 229 Chr. 7 - 44831575 - 44831834 [+] on Build GRCm38
          GenBankmRNA BB041423.1 - 1 262 Chr. 7 - 44831551 - 44831834 [+] on Build GRCm38
          GenBankmRNA BB090475.1 - 1 244 Chr. 7 - 44831559 - 44831834 [+] on Build GRCm38
          GenBankmRNA AV306808.1 - 1 295 Chr. 7 - 44831550 - 44831833 [+] on Build GRCm38
          GenBankmRNA BB771243.1 - 1 498 Chr. 7 - 44831305 - 44831834 [+] on Build GRCm38
          GenBankmRNA BY576585.1 - 1 364 Chr. 7 - 44831440 - 44831834 [+] on Build GRCm38
          GenBankmRNA BB147144.1 - 1 299 Chr. 7 - 44831518 - 44831834 [+] on Build GRCm38
          GenBankmRNA BY579761.1 - 1 400 Chr. 7 - 44831403 - 44831830 [+] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline