Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAGT1 SIRNA Assays ›
          Gene Symbol:
          Magt1
          Gene Name:
          magnesium transporter 1
          Gene Aliases:
          2410001C15Rik, 2610529C04Rik, 2810482I07Rik, IAG2, IAP
          Chromosome Location:
          Chr.X: 105968084 - 106011899 on Build GRCm38
          UniGene ID:
          Mm.275943
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n431436

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Magt1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK013434.1 - 1 1100
          AV024949.2 - 1 724
          BC023242.1 - 1 79
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY671914.1 - 1 95 Chr. X - 105968087 - 105968474 [-] on Build GRCm38
          GenBankmRNA CF751765.1 - 1 430 Chr. X - 105968084 - 105968809 [-] on Build GRCm38
          GenBankmRNA EH094033.1 - 1 146 Chr. X - 105968204 - 105968510 [-] on Build GRCm38
          GenBankmRNA BX526148.1 - 1 419 Chr. X - 105968264 - 105968798 [-] on Build GRCm38
          GenBankmRNA BE448605.1 - 1 470 Chr. X - 105968362 - 105968849 [-] on Build GRCm38
          GenBankmRNA BY663856.1 - 1 61 Chr. X - 105968087 - 105968440 [-] on Build GRCm38
          GenBankmRNA BI456382.1 - 1 431 Chr. X - 105968079 - 105968810 [-] on Build GRCm38
          GenBankmRNA BY408470.1 - 1 79 Chr. X - 105968084 - 105968457 [-] on Build GRCm38
          GenBankmRNA BF661827.1 - 1 232 Chr. X - 105968182 - 105968611 [-] on Build GRCm38
          GenBankmRNA BB728184.1 - 1 149 Chr. X - 105968087 - 105968529 [-] on Build GRCm38
          GenBankmRNA BY596809.1 - 1 172 Chr. X - 105968092 - 105968551 [-] on Build GRCm38
          GenBankmRNA BY669398.1 - 1 86 Chr. X - 105968085 - 105968465 [-] on Build GRCm38
          GenBankmRNA AL362432.1 - 1 131 Chr. X - 105968116 - 105968499 [-] on Build GRCm38
          GenBankmRNA BF452675.1 - 1 130 Chr. X - 105968165 - 105968509 [-] on Build GRCm38
          GenBankmRNA BY409966.1 - 1 95 Chr. X - 105968087 - 105968474 [-] on Build GRCm38
          GenBankmRNA CJ306179.1 - 1 118 Chr. X - 105968087 - 105968497 [-] on Build GRCm38
          GenBankmRNA BB812812.1 - 1 106 Chr. X - 105968087 - 105968485 [-] on Build GRCm38
          GenBankmRNA BG091025.1 - 1 231 Chr. X - 105968266 - 105968610 [-] on Build GRCm38
          GenBankmRNA BY578955.1 - 1 114 Chr. X - 105968087 - 105968493 [-] on Build GRCm38
          GenBankmRNA BY667881.1 - 1 99 Chr. X - 105968087 - 105968474 [-] on Build GRCm38
          GenBankmRNA CK636450.1 - 1 334 Chr. X - 105968087 - 105968713 [-] on Build GRCm38
          GenBankmRNA BG091474.1 - 1 231 Chr. X - 105968241 - 105968610 [-] on Build GRCm38
          GenBankmRNA CJ322597.1 - 1 113 Chr. X - 105968087 - 105968492 [-] on Build GRCm38
          GenBankmRNA BB780645.1 - 1 163 Chr. X - 105968092 - 105968542 [-] on Build GRCm38
          GenBankmRNA BB768380.1 - 1 246 Chr. X - 105968087 - 105968623 [-] on Build GRCm38
          GenBankmRNA BY406381.1 - 1 76 Chr. X - 105968090 - 105968454 [-] on Build GRCm38
          GenBankmRNA BY606319.1 - 1 78 Chr. X - 105968087 - 105968457 [-] on Build GRCm38
          GenBankmRNA AA245303.1 - 1 203 Chr. X - 105968134 - 105968582 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0