Search
Search

VPS13A-AS1| Gene Symbol: VPS13A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026668.2 | 1, 2 | 293 | |||
| GenBank mRNA | DB509755.1 | - | 1, 2 | 299 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000415172.1 | - | 1, 2 | 370 | Chr. 9 - 77176756 - 77177994 [-] on Build GRCh38 | |