Search
Search

DLG5-AS1| Gene Symbol: DLG5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024585.1 | 1 | 613 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000449852.1 | - | 1 | 53 | Chr. 10 - 77927372 - 77929824 [+] on Build GRCh38 | |