Search
Search

PSMB8-AS1| Gene Symbol: PSMB8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037173.1 | 3 | 945 | |||
| NR_037174.1 | 2 | 807 | ||||
| NR_037175.1 | 3 | 780 | ||||
| NR_037176.1 | 2 | 719 | ||||
| GenBank mRNA | BI906764.1 | - | 2 | 672 | ||
| BU539272.1 | - | 2 | 756 | |||