Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEG3 SIRNA Assays ›
          Gene Symbol:
          MEG3
          Gene Name:
          maternally expressed 3 (non-protein coding)
          Gene Aliases:
          FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
          Chromosome Location:
          Chr.14: 100826108 - 100861026 on Build GRCh38
          UniGene ID:
          Hs.654863
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n503814

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002766.2
          5 1092
          NR_003530.2
          7 1342
          NR_003531.3
          6 1209
          NR_033358.1
          6 1222
          NR_033359.1
          6 1201
          NR_033360.1
          5 1108
          NR_046464.1
          6 1156
          NR_046465.2
          5 1130
          NR_046466.1
          4 978
          NR_046467.1
          6 1260
          NR_046469.1
          5 1122
          NR_046470.2
          6 1252
          NR_046471.1
          4 1016
          NR_046472.1
          5 1146
          NR_046473.1
          5 1092
          GenBank mRNA AB032607.1 -
          5 1078
          AF052114.1 -
          3 834
          AJ413186.1 -
          2 2355
          AK055725.1 -
          2 2366
          AK057522.1 -
          4 1228
          AK092504.1 -
          3 1780
          AK092707.1 -
          2 1648
          AK127864.1 -
          2 2955
          AV704361.1 -
          1 16
          AY314975.1 - LIT3321
          MEG3
          6 1208
          BC036882.1 -
          3 217
          BC062783.1 -
          2 544
          BC092509.1 -
          3 287
          GQ183494.1 -
          5 1100
          GQ183495.1 -
          6 1148
          GQ183496.1 -
          5 1123
          GQ183497.1 -
          4 970
          GQ183498.1 -
          6 1252
          GQ183499.1 -
          6 1215
          GQ183500.1 -
          5 1114
          GQ183501.1 -
          6 1245
          GQ183502.1 -
          4 1008
          GQ183503.1 -
          5 1138
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GENCODE ENST00000451743.6 - 5 1074 Chr. 14 - 100826126 - 100861031 [+] on Build GRCh38
          GenBankmRNA CD513101.1 - 2 169 Chr. 14 - 100835754 - 100861026 [+] on Build GRCh38
          GenBankmRNA BF932705.1 - 336 Chr. - - [-] on Build
          GenBankmRNA BU075874.1 - 2 357 Chr. 14 - 100835573 - 100860746 [+] on Build GRCh38
          GenBankmRNA R81006.1 - 149 Chr. - - [-] on Build
          GenBankmRNA AV701976.1 - 3 242 Chr. 14 - 100835307 - 100860739 [+] on Build GRCh38
          GenBankmRNA BX426730.2 - 2 86 Chr. 14 - 100831491 - 100845533 [+] on Build GRCh38
          GenBankmRNA BX453065.2 - 140 Chr. - - [-] on Build
          GenBankmRNA BX346951.1 - 3 414 Chr. 14 - 100829508 - 100860951 [+] on Build GRCh38
          GenBankmRNA BX408784.2 - 3 331 Chr. 14 - 100829579 - 100861025 [+] on Build GRCh38
          GENCODE ENST00000554639.5 - 6 1209 Chr. 14 - 100826132 - 100861026 [+] on Build GRCh38
          GenBankmRNA AI174720.1 - 3 510 Chr. 14 - 100829397 - 100836304 [+] on Build GRCh38
          GenBankmRNA BM929634.1 - 3 229 Chr. 14 - 100835728 - 100861002 [+] on Build GRCh38
          GenBankmRNA BU786542.1 - 1 407 Chr. 14 - 100835806 - 100860767 [+] on Build GRCh38
          GenBankmRNA BM699430.1 - 4 368 Chr. 14 - 100831460 - 100860756 [+] on Build GRCh38
          GENCODE ENST00000412736.6 - 6 1161 Chr. 14 - 100826148 - 100860977 [+] on Build GRCh38
          GenBankmRNA H59650.1 - 286 Chr. - - [-] on Build
          GenBankmRNA BX408697.2 - 3 282 Chr. 14 - 100829658 - 100861025 [+] on Build GRCh38
          GENCODE ENST00000429159.6 - 5 1108 Chr. 14 - 100826108 - 100861023 [+] on Build GRCh38
          GenBankmRNA BX367746.1 - 3 499 Chr. 14 - 100829409 - 100860888 [+] on Build GRCh38
          GENCODE ENST00000398474.7 - 4 1223 Chr. 14 - 100830605 - 100861021 [+] on Build GRCh38
          GENCODE ENST00000555928.5 - 3 217 Chr. 14 - 100831481 - 100861026 [+] on Build GRCh38
          GenBankmRNA BU783773.1 - 2 130 Chr. 14 - 100831456 - 100860960 [+] on Build GRCh38
          GenBankmRNA DA361098.1 - 3 271 Chr. 14 - 100829636 - 100845997 [+] on Build GRCh38
          GenBankmRNA AV723467.1 - 3 247 Chr. 14 - 100829658 - 100860731 [+] on Build GRCh38
          GenBankmRNA CB154926.1 - 3 339 Chr. 14 - 100829568 - 100860741 [+] on Build GRCh38
          GenBankmRNA AA029246.1 - 2 129 Chr. 14 - 100831448 - 100860775 [+] on Build GRCh38
          GenBankmRNA BX453064.2 - 2 141 Chr. 14 - 100831437 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX422168.2 - 3 564 Chr. 14 - 100829359 - 100860947 [+] on Build GRCh38
          GenBankmRNA BX408189.1 - 3 445 Chr. 14 - 100829460 - 100860925 [+] on Build GRCh38
          GenBankmRNA AU309923.1 - 3 268 Chr. 14 - 100829639 - 100846016 [+] on Build GRCh38
          GenBankmRNA AI459255.1 - 2 120 Chr. 14 - 100835999 - 100860852 [+] on Build GRCh38
          GenBankmRNA BX423733.2 - 2 87 Chr. 14 - 100831491 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX450687.2 - 3 323 Chr. 14 - 100829584 - 100860988 [+] on Build GRCh38
          GenBankmRNA BI913162.1 - 2 140 Chr. 14 - 100831438 - 100861007 [+] on Build GRCh38
          GenBankmRNA BU579722.1 - 2 100 Chr. 14 - 100831478 - 100860862 [+] on Build GRCh38
          GENCODE ENST00000455531.1 - 1 2932 Chr. 14 - 100833278 - 100847301 [+] on Build GRCh38
          GenBankmRNA BU070003.1 - 1 169 Chr. 14 - 100836048 - 100860862 [+] on Build GRCh38
          GENCODE ENST00000556736.5 - 6 1204 Chr. 14 - 100826126 - 100861008 [+] on Build GRCh38
          GenBankmRNA BX326323.2 - 3 293 Chr. 14 - 100829630 - 100861050 [+] on Build GRCh38
          GenBankmRNA H48704.1 - 2 85 Chr. 14 - 100835837 - 100860825 [+] on Build GRCh38
          GenBankmRNA BY796127.2 - 3 461 Chr. 14 - 100829446 - 100845508 [+] on Build GRCh38
          GenBankmRNA BG703541.1 - 3 292 Chr. 14 - 100829615 - 100846219 [+] on Build GRCh38
          GenBankmRNA CA943189.1 - 3 386 Chr. 14 - 100829522 - 100845520 [+] on Build GRCh38
          GenBankmRNA BU952631.1 - 2 299 Chr. 14 - 100835624 - 100860744 [+] on Build GRCh38
          GenBankmRNA BU070059.1 - 1 169 Chr. 14 - 100836048 - 100860906 [+] on Build GRCh38
          GenBankmRNA BX368158.1 - 3 534 Chr. 14 - 100829371 - 100860859 [+] on Build GRCh38
          GenBankmRNA BU787606.1 - 2 100 Chr. 14 - 100831478 - 100861005 [+] on Build GRCh38
          GENCODE ENST00000398518.6 - 6 1203 Chr. 14 - 100826127 - 100860994 [+] on Build GRCh38
          GenBankmRNA BQ549202.1 - 1 68 Chr. 14 - 100836162 - 100860895 [+] on Build GRCh38
          GenBankmRNA CR741469.1 - 1 133 Chr. 14 - 100836077 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX368151.1 - 5 443 Chr. 14 - 100829640 - 100860985 [+] on Build GRCh38
          GENCODE ENST00000524035.5 - 2 2355 Chr. 14 - 100833764 - 100861021 [+] on Build GRCh38
          GenBankmRNA BX450276.1 - 3 423 Chr. 14 - 100829484 - 100846142 [+] on Build GRCh38
          GenBankmRNA BX408219.1 - 3 399 Chr. 14 - 100829507 - 100860967 [+] on Build GRCh38
          GENCODE ENST00000521404.5 - 4 960 Chr. 14 - 100826126 - 100860989 [+] on Build GRCh38
          GenBankmRNA BX428872.2 - 2 141 Chr. 14 - 100831437 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX408790.2 - 3 238 Chr. 14 - 100829668 - 100860997 [+] on Build GRCh38
          GenBankmRNA BU070170.1 - 1 169 Chr. 14 - 100836048 - 100860821 [+] on Build GRCh38
          GenBankmRNA BI916294.1 - 3 217 Chr. 14 - 100831481 - 100860983 [+] on Build GRCh38
          GenBankmRNA AA009857.1 - 2 120 Chr. 14 - 100835805 - 100860934 [+] on Build GRCh38
          GenBankmRNA DA083332.1 - 3 394 Chr. 14 - 100829513 - 100836289 [+] on Build GRCh38
          GenBankmRNA BU075431.1 - 3 250 Chr. 14 - 100829679 - 100860862 [+] on Build GRCh38
          GenBankmRNA BM875701.1 - 2 76 Chr. 14 - 100835854 - 100860960 [+] on Build GRCh38
          GenBankmRNA BX429998.2 - 3 217 Chr. 14 - 100829691 - 100846355 [+] on Build GRCh38
          GenBankmRNA BQ268259.1 - 2 144 Chr. 14 - 100835786 - 100860745 [+] on Build GRCh38
          GenBankmRNA BQ669975.1 - 152 Chr. - - [-] on Build
          GENCODE ENST00000521812.1 - 2 621 Chr. 14 - 100832064 - 100836223 [+] on Build GRCh38
          GenBankmRNA AV727351.1 - 4 528 Chr. 14 - 100829325 - 100845483 [+] on Build GRCh38
          GenBankmRNA AI133465.1 - 2 201 Chr. 14 - 100835722 - 100861003 [+] on Build GRCh38
          GenBankmRNA BX429896.2 - 3 282 Chr. 14 - 100829639 - 100846333 [+] on Build GRCh38
          GenBankmRNA CA949980.1 - 2 137 Chr. 14 - 100832548 - 100860960 [+] on Build GRCh38
          GENCODE ENST00000522618.1 - 1 490 Chr. 14 - 100835720 - 100860987 [+] on Build GRCh38
          GenBankmRNA AV753036.1 - 433 Chr. - - [-] on Build
          GenBankmRNA N56264.1 - 2 110 Chr. 14 - 100831464 - 100845534 [+] on Build GRCh38
          GenBankmRNA BX347225.2 - 5 611 Chr. 14 - 100829557 - 100836299 [+] on Build GRCh38
          GENCODE ENST00000398461.5 - 2 2954 Chr. 14 - 100832969 - 100861021 [+] on Build GRCh38
          GenBankmRNA BU079357.1 - 2 116 Chr. 14 - 100836004 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX408226.1 - 3 416 Chr. 14 - 100829491 - 100860979 [+] on Build GRCh38
          GenBankmRNA BQ478473.1 - 2 72 Chr. 14 - 100835852 - 100860906 [+] on Build GRCh38
          GenBankmRNA BI524001.1 - 79 Chr. - - [-] on Build
          GenBankmRNA BX429869.2 - 3 279 Chr. 14 - 100829629 - 100861023 [+] on Build GRCh38
          GENCODE ENST00000423456.5 - 7 1324 Chr. 14 - 100826126 - 100860970 [+] on Build GRCh38
          GenBankmRNA BX367745.1 - 2 515 Chr. 14 - 100835613 - 100860871 [+] on Build GRCh38
          GenBankmRNA BU580862.1 - 3 365 Chr. 14 - 100829558 - 100860756 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0