Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEG3 SIRNA Assays ›
          Gene Symbol:
          MEG3
          Gene Name:
          maternally expressed 3 (non-protein coding)
          Gene Aliases:
          FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
          Chromosome Location:
          Chr.14: 100826108 - 100861026 on Build GRCh38
          UniGene ID:
          Hs.654863
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n503815

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002766.2
          7 1527
          NR_003530.2
          9 1777
          NR_003531.3
          8 1644
          NR_033358.1
          8 1657
          NR_033359.1
          8 1636
          NR_033360.1
          7 1543
          NR_046464.1
          8 1591
          NR_046465.2
          7 1565
          NR_046466.1
          6 1413
          NR_046467.1
          8 1695
          NR_046469.1
          7 1557
          NR_046470.2
          8 1687
          NR_046471.1
          6 1451
          NR_046472.1
          7 1581
          GenBank mRNA AB032607.1 -
          7 1511
          AJ413186.1 -
          4 2790
          AK055725.1 -
          4 2801
          AK057522.1 -
          6 1663
          AK092504.1 -
          5 2215
          AK092707.1 -
          4 2083
          AK127864.1 -
          4 3390
          AY314975.1 - LIT3321
          MEG3
          8 1643
          BC036882.1 -
          5 652
          BC062783.1 -
          4 979
          GQ183494.1 -
          7 1535
          GQ183495.1 -
          8 1583
          GQ183496.1 -
          7 1558
          GQ183497.1 -
          6 1405
          GQ183498.1 -
          8 1687
          GQ183499.1 -
          8 1650
          GQ183500.1 -
          7 1549
          GQ183501.1 -
          8 1680
          GQ183502.1 -
          6 1443
          GQ183503.1 -
          7 1573
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GENCODE ENST00000451743.6 - 7 1509 Chr. 14 - 100826126 - 100861031 [+] on Build GRCh38
          GenBankmRNA BI913162.1 - 4 575 Chr. 14 - 100831438 - 100861007 [+] on Build GRCh38
          GenBankmRNA CD513101.1 - 4 604 Chr. 14 - 100835754 - 100861026 [+] on Build GRCh38
          GenBankmRNA DB514072.1 - 3 385 Chr. 14 - 100836260 - 100861026 [+] on Build GRCh38
          GENCODE ENST00000556736.5 - 8 1639 Chr. 14 - 100826126 - 100861008 [+] on Build GRCh38
          GENCODE ENST00000398460.3 - 2 3322 Chr. 14 - 100857245 - 100861018 [+] on Build GRCh38
          GENCODE ENST00000554639.5 - 8 1644 Chr. 14 - 100826132 - 100861026 [+] on Build GRCh38
          GenBankmRNA DB573877.1 - 3 404 Chr. 14 - 100836241 - 100861021 [+] on Build GRCh38
          GenBankmRNA BM929634.1 - 5 664 Chr. 14 - 100835728 - 100861002 [+] on Build GRCh38
          GenBankmRNA BU787606.1 - 4 535 Chr. 14 - 100831478 - 100861005 [+] on Build GRCh38
          GenBankmRNA DB547298.1 - 3 362 Chr. 14 - 100836283 - 100861021 [+] on Build GRCh38
          GENCODE ENST00000398518.6 - 8 1638 Chr. 14 - 100826127 - 100860994 [+] on Build GRCh38
          GenBankmRNA DB541803.1 - 3 411 Chr. 14 - 100836234 - 100861021 [+] on Build GRCh38
          GenBankmRNA DB540320.1 - 3 430 Chr. 14 - 100836215 - 100861021 [+] on Build GRCh38
          GenBankmRNA BM512552.1 - 2 343 Chr. 14 - 100845466 - 100861026 [+] on Build GRCh38
          GENCODE ENST00000412736.6 - 8 1596 Chr. 14 - 100826148 - 100860977 [+] on Build GRCh38
          GenBankmRNA DB573200.1 - 3 420 Chr. 14 - 100836226 - 100861021 [+] on Build GRCh38
          GENCODE ENST00000429159.6 - 7 1543 Chr. 14 - 100826108 - 100861023 [+] on Build GRCh38
          GENCODE ENST00000398474.7 - 6 1658 Chr. 14 - 100830605 - 100861021 [+] on Build GRCh38
          GenBankmRNA DB562479.1 - 2 356 Chr. 14 - 100845471 - 100861021 [+] on Build GRCh38
          GenBankmRNA DB541166.1 - 3 404 Chr. 14 - 100836242 - 100861021 [+] on Build GRCh38
          GenBankmRNA CR741469.1 - 3 568 Chr. 14 - 100836077 - 100861026 [+] on Build GRCh38
          GENCODE ENST00000524035.5 - 4 2790 Chr. 14 - 100833764 - 100861021 [+] on Build GRCh38
          GenBankmRNA AI299753.1 - 2 327 Chr. 14 - 100845475 - 100861007 [+] on Build GRCh38
          GENCODE ENST00000521404.5 - 6 1395 Chr. 14 - 100826126 - 100860989 [+] on Build GRCh38
          GenBankmRNA DB557904.1 - 3 376 Chr. 14 - 100836268 - 100861021 [+] on Build GRCh38
          GENCODE ENST00000555928.5 - 5 652 Chr. 14 - 100831481 - 100861026 [+] on Build GRCh38
          GenBankmRNA BX428872.2 - 4 575 Chr. 14 - 100831437 - 100861026 [+] on Build GRCh38
          GenBankmRNA BI916294.1 - 5 666 Chr. 14 - 100831481 - 100860983 [+] on Build GRCh38
          GenBankmRNA DB535072.1 - 3 382 Chr. 14 - 100836263 - 100861021 [+] on Build GRCh38
          GenBankmRNA AI133465.1 - 4 636 Chr. 14 - 100835722 - 100861003 [+] on Build GRCh38
          GENCODE ENST00000522618.1 - 3 925 Chr. 14 - 100835720 - 100860987 [+] on Build GRCh38
          GenBankmRNA BX453064.2 - 4 576 Chr. 14 - 100831437 - 100861026 [+] on Build GRCh38
          GENCODE ENST00000398461.5 - 4 3389 Chr. 14 - 100832969 - 100861021 [+] on Build GRCh38
          GenBankmRNA DB575067.1 - 3 398 Chr. 14 - 100836247 - 100861021 [+] on Build GRCh38
          GenBankmRNA BU079357.1 - 4 551 Chr. 14 - 100836004 - 100861026 [+] on Build GRCh38
          GenBankmRNA DB514120.1 - 3 400 Chr. 14 - 100836244 - 100861021 [+] on Build GRCh38
          GenBankmRNA DB543949.1 - 3 391 Chr. 14 - 100836254 - 100861021 [+] on Build GRCh38
          GenBankmRNA DB551840.1 - 3 404 Chr. 14 - 100836241 - 100861021 [+] on Build GRCh38
          GenBankmRNA BI524001.1 - 514 Chr. - - [-] on Build
          GenBankmRNA DB546570.1 - 3 365 Chr. 14 - 100836280 - 100861021 [+] on Build GRCh38
          GenBankmRNA DB567079.1 - 3 406 Chr. 14 - 100836239 - 100861021 [+] on Build GRCh38
          GENCODE ENST00000423456.5 - 9 1759 Chr. 14 - 100826126 - 100860970 [+] on Build GRCh38
          GenBankmRNA BP395862.1 - 3 431 Chr. 14 - 100836214 - 100861022 [+] on Build GRCh38
          GenBankmRNA DB561566.1 - 2 358 Chr. 14 - 100845444 - 100861021 [+] on Build GRCh38
          GenBankmRNA BM511346.1 - 2 343 Chr. 14 - 100845466 - 100861026 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4pjxj:80/100.66.79.163:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0