Search
Search

HTR5A-AS1| Gene Symbol: HTR5A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038945.1 | 2 | 2006 | |||
| GenBank mRNA | BC031272.1 | - | 2 | 1978 | ||
| BC045795.1 | - | 2 | 1981 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000395731.5 | - | 2 | 2006 | Chr. 7 - 155067067 - 155071557 [-] on Build GRCh38 | |