Search
Search

CCNT2-AS1| Gene Symbol: CCNT2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036549.1 | 5 | 704 | |||
| GenBank mRNA | AK303295.1 | - | 5 | 704 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000413962.5 | - | 3 | 620 | Chr. 2 - 134867318 - 134918678 [-] on Build GRCh38 | |