Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JPX SIRNA Assays ›
          Gene Symbol:
          JPX
          Gene Name:
          JPX transcript, XIST activator (non-protein coding)
          Gene Aliases:
          DCBALD06, ENOX, LINC00183, NCRNA00183
          Chromosome Location:
          Chr.X: 73944324 - 74070384 on Build GRCh38
          UniGene ID:
          Hs.648316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n504763

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JPX
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024582.1
          1 59
          GenBank mRNA AK056172.1 -
          1 41
          AV714079.1 - LIT1011
          Jpx
          44
          BC071776.1 -
          1 55
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA979750.1 - 1 32 Chr. X - 73944351 - 73947352 [+] on Build GRCh38
          GenBankmRNA BG504771.1 - 1 38 Chr. X - 73944346 - 73947528 [+] on Build GRCh38
          GenBankmRNA BP872745.1 - 1 38 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP871836.1 - 1 37 Chr. X - 73944346 - 73947297 [+] on Build GRCh38
          GenBankmRNA CN270680.1 - 1 24 Chr. X - 73944359 - 73999372 [+] on Build GRCh38
          GenBankmRNA AW963219.1 - 31 Chr. - - [-] on Build
          GenBankmRNA BU934969.1 - 1 39 Chr. X - 73944346 - 73999254 [+] on Build GRCh38
          GenBankmRNA AL711317.1 - 1 59 Chr. X - 73944324 - 73999368 [+] on Build GRCh38
          GenBankmRNA BG612854.1 - 1 40 Chr. X - 73944343 - 73999478 [+] on Build GRCh38
          GenBankmRNA CB993733.1 - 68 Chr. - - [-] on Build
          GenBankmRNA HY065667.1 - 1 45 Chr. X - 73944340 - 74010499 [+] on Build GRCh38
          GenBankmRNA BP872499.1 - 1 59 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP873595.1 - 1 41 Chr. X - 73944342 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP872256.1 - 1 38 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA DB255033.1 - 1 44 Chr. X - 73944339 - 73947366 [+] on Build GRCh38
          GenBankmRNA CF129475.1 - 1 24 Chr. X - 73944359 - 74004553 [+] on Build GRCh38
          GenBankmRNA DA363254.1 - 1 57 Chr. X - 73944326 - 73997077 [+] on Build GRCh38
          GenBankmRNA DA106100.1 - 1 41 Chr. X - 73944342 - 74004417 [+] on Build GRCh38
          GENCODE ENST00000414209.5 - 1 41 Chr. X - 73944342 - 74000491 [+] on Build GRCh38
          GenBankmRNA CN263834.1 - 1 27 Chr. X - 73944356 - 73999314 [+] on Build GRCh38
          GenBankmRNA HY103315.1 - 1 41 Chr. X - 73944342 - 73947268 [+] on Build GRCh38
          GENCODE ENST00000453317.1 - 1 37 Chr. X - 73944346 - 73944946 [+] on Build GRCh38
          GenBankmRNA BM810643.1 - 14 Chr. - - [-] on Build
          GenBankmRNA BU164379.1 - 1 34 Chr. X - 73944349 - 73998832 [+] on Build GRCh38
          GenBankmRNA BP871739.1 - 1 38 Chr. X - 73944345 - 74004345 [+] on Build GRCh38
          GenBankmRNA BP872588.1 - 1 38 Chr. X - 73944345 - 74004345 [+] on Build GRCh38
          GenBankmRNA BP871919.1 - 1 39 Chr. X - 73944344 - 73999613 [+] on Build GRCh38
          GenBankmRNA DA841100.1 - 1 38 Chr. X - 73944345 - 73947361 [+] on Build GRCh38
          GenBankmRNA BG613915.1 - 1 42 Chr. X - 73944342 - 73999860 [+] on Build GRCh38
          GenBankmRNA AL708895.1 - 1 46 Chr. X - 73944338 - 73947333 [+] on Build GRCh38
          GenBankmRNA BU537716.1 - 1 53 Chr. X - 73944333 - 73947334 [+] on Build GRCh38
          GenBankmRNA BP871940.1 - 1 38 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA DA331725.1 - 1 41 Chr. X - 73944342 - 74004540 [+] on Build GRCh38
          GenBankmRNA BP873466.1 - 1 59 Chr. X - 73944324 - 73999111 [+] on Build GRCh38
          GENCODE ENST00000602294.5 - 1 59 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA BP871783.1 - 1 38 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA HY172225.1 - 1 42 Chr. X - 73944342 - 73999019 [+] on Build GRCh38
          GenBankmRNA HY088843.1 - 1 45 Chr. X - 73944340 - 74010567 [+] on Build GRCh38
          GenBankmRNA BP872277.1 - 1 59 Chr. X - 73944324 - 73999592 [+] on Build GRCh38
          GenBankmRNA BG435566.1 - 1 56 Chr. X - 73944328 - 74069325 [+] on Build GRCh38
          GenBankmRNA DA103086.1 - 1 40 Chr. X - 73944343 - 74004499 [+] on Build GRCh38
          GenBankmRNA BU589636.1 - 1 40 Chr. X - 73944346 - 73944948 [+] on Build GRCh38
          GenBankmRNA DA427047.1 - 1 57 Chr. X - 73944326 - 73947342 [+] on Build GRCh38
          GenBankmRNA DB240432.1 - 1 37 Chr. X - 73944346 - 73947385 [+] on Build GRCh38
          GenBankmRNA CB991754.1 - 68 Chr. - - [-] on Build
          GenBankmRNA DA679611.1 - 1 41 Chr. X - 73944342 - 73999105 [+] on Build GRCh38
          GenBankmRNA DA881370.1 - 1 36 Chr. X - 73944347 - 73999014 [+] on Build GRCh38
          GenBankmRNA BP873513.1 - 1 38 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA DA731241.1 - 1 57 Chr. X - 73944326 - 73998845 [+] on Build GRCh38
          GenBankmRNA BP872419.1 - 1 59 Chr. X - 73944324 - 74004351 [+] on Build GRCh38
          GenBankmRNA DA700446.1 - 1 41 Chr. X - 73944342 - 73947318 [+] on Build GRCh38
          GenBankmRNA DB250056.1 - 1 37 Chr. X - 73944346 - 73999096 [+] on Build GRCh38
          GenBankmRNA BF686802.1 - 22 Chr. - - [-] on Build
          GenBankmRNA BG427198.1 - 1 42 Chr. X - 73944342 - 73995436 [+] on Build GRCh38
          GenBankmRNA DW450219.1 - 1 54 Chr. X - 73944342 - 73944645 [+] on Build GRCh38
          GenBankmRNA DW466961.1 - 1 56 Chr. X - 73944340 - 73994872 [+] on Build GRCh38
          GenBankmRNA BP871862.1 - 1 43 Chr. X - 73944340 - 73947374 [+] on Build GRCh38
          GenBankmRNA BP873504.1 - 1 35 Chr. X - 73944356 - 74004351 [+] on Build GRCh38
          GENCODE ENST00000602772.5 - 1 55 Chr. X - 73944328 - 74070408 [+] on Build GRCh38
          GenBankmRNA BP872587.1 - 1 39 Chr. X - 73944344 - 74004351 [+] on Build GRCh38
          GenBankmRNA HY081032.1 - 1 44 Chr. X - 73944339 - 73947244 [+] on Build GRCh38
          GENCODE ENST00000602938.5 - 1 59 Chr. X - 73944324 - 73999114 [+] on Build GRCh38
          GenBankmRNA BP873185.1 - 1 39 Chr. X - 73944344 - 74004355 [+] on Build GRCh38
          GenBankmRNA BU599790.1 - 1 44 Chr. X - 73944340 - 74004542 [+] on Build GRCh38
          GenBankmRNA BP872964.1 - 1 38 Chr. X - 73944345 - 74004351 [+] on Build GRCh38
          GenBankmRNA BF797301.1 - 1 26 Chr. X - 73944357 - 73999368 [+] on Build GRCh38
          GenBankmRNA BP872836.1 - 1 39 Chr. X - 73944344 - 74004351 [+] on Build GRCh38
          GenBankmRNA DB210310.1 - 1 51 Chr. X - 73944332 - 73999117 [+] on Build GRCh38
          GenBankmRNA DA711535.1 - 1 42 Chr. X - 73944342 - 73947373 [+] on Build GRCh38
          GenBankmRNA CB998281.1 - 68 Chr. - - [-] on Build
          GENCODE ENST00000602920.5 - 1 59 Chr. X - 73944324 - 74004551 [+] on Build GRCh38
          GenBankmRNA BP872490.1 - 1 39 Chr. X - 73944344 - 74004345 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0