Search
Search

FGF13-AS1| Gene Symbol: FGF13-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038405.1 | 3 | 634 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000438238.1 | - | 3 | 634 | Chr. X - 138712107 - 138716617 [+] on Build GRCh38 | |