Search
Search

CLYBL-AS2| Gene Symbol: CLYBL-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046526.1 | 1 | 212 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000430125.1 | - | 1 | 212 | Chr. 13 - 99690081 - 99690971 [-] on Build GRCh38 | |