Search
Search

ELOVL2-AS1| Gene Symbol: ELOVL2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038962.1 | 1, 2 | 280 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456616.1 | - | 1, 2 | 278 | Chr. 6 - 11044693 - 11078226 [+] on Build GRCh38 | |