Search
Search

ELOVL2-AS1| Gene Symbol: ELOVL2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038962.1 | 3 | 444 | |||
| NR_038963.1 | 3 | 236 | ||||
| NR_038964.1 | 5 | 438 | ||||
| GenBank mRNA | BC038757.1 | - | 3 | 444 | ||
| BC044599.1 | - | 3 | 247 | |||
| BI828717.1 | - | 5 | 439 | |||
| DN918479.1 | - | 3 | 214 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456616.1 | - | 3 | 442 | Chr. 6 - 11044693 - 11078226 [+] on Build GRCh38 | |