Search
Search

VAV3-AS1| Gene Symbol: VAV3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046653.1 | 3 | 525 | |||
| GenBank mRNA | BG193383.1 | - | 1 | 333 | ||
| CR744964.1 | - | 3 | 525 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000438318.1 | - | 3 | 525 | Chr. 1 - 107964443 - 107994607 [+] on Build GRCh38 | |