Search
Search

FOXD3-AS1| Gene Symbol: FOXD3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121634.1 | 4 | 797 | |||
| NR_121635.1 | 3 | 381 | ||||
| NR_121636.1 | 3 | 423 | ||||
| NR_121637.1 | 3 | 325 | ||||
| GenBank mRNA | KF925349.1 | - | 3 | 325 | ||
| KF925350.1 | - | 3 | 330 | |||
| KF925351.1 | - | 3 | 381 | |||
| KF925352.1 | - | 4 | 775 | |||
| KF925353.1 | - | 3 | 266 | |||
| KF925354.1 | - | 2 | 304 | |||