Search
Search

ATP11A-AS1| Gene Symbol: ATP11A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046661.1 | 2 | 1430 | |||
| GenBank mRNA | BC041346.1 | - | 2 | 1390 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000446442.1 | - | 2 | 1392 | Chr. 13 - 112745449 - 112754693 [-] on Build GRCh38 | |