Search
Search

STARD13-AS| Gene Symbol: STARD13-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046693.1 | 2 | 195 | |||
| GenBank mRNA | BX103803.1 | - | 2 | 195 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000588660.5 | - | 2 | 196 | Chr. 13 - 33277553 - 33281334 [+] on Build GRCh38 | |