Search
Search

PKN2-AS1| Gene Symbol: PKN2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110682.1 | 4 | 2322 | |||
| GenBank mRNA | AK123834.1 | - | 4 | 2312 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000458097.5 | - | 4 | 2322 | Chr. 1 - 88537513 - 88685204 [-] on Build GRCh38 | |