Search
Search

A2M-AS1| Gene Symbol: A2M-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026971.1 | 3 | 1617 | |||
| GenBank mRNA | AK056852.1 | - | 3 | 1617 | ||
| AK096375.1 | - | 1 | 1494 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499762.2 | - | 3 | 1617 | Chr. 12 - 9065177 - 9068055 [+] on Build GRCh38 | |