Search
Search

LRP4-AS1| Gene Symbol: LRP4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038909.1 | 3 | 875 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000502049.2 | - | 3 | 859 | Chr. 11 - 46846427 - 46874396 [+] on Build GRCh38 | |