Search
Search

UGDH-AS1| Gene Symbol: UGDH-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047679.1 | 5 | 1486 | |||
| GenBank mRNA | AK056558.1 | - | 5 | 1306 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000504032.1 | - | 5 | 1306 | Chr. 4 - 39528019 - 39594707 [+] on Build GRCh38 | |