Search
Search

LOC101929528| Gene Symbol: LOC101929528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105005.1 | 8 | 1575 | |||
| GenBank mRNA | BC032030.1 | - | 8 | 1575 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000519981.5 | - | 8 | 1575 | Chr. 8 - 58255771 - 58272101 [-] on Build GRCh38 | |