Search
Search

ZFHX4-AS1| Gene Symbol: ZFHX4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024360.1 | 4 | 1614 | |||
| GenBank mRNA | BC037827.1 | - | 4 | 1622 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000518143.1 | - | 4 | 1617 | Chr. 8 - 76610881 - 76683278 [-] on Build GRCh38 | |