Search
Search

LOC100287896| Gene Symbol: LOC100287896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319240.1 | NP_001306169.1 | 2 | 601 | ||
| XM_017017055.1 | XP_016872544.1 | 2 | 89 | |||
| GenBank mRNA | BC019340.1 | - | 2 | 546 | 3051084 | |
| BX647206.1 | - | 2 | 190 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000526036.1 | - | 2 | 620 | Chr. 11 - 74493366 - 74498533 [+] on Build GRCh38 | |