Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEG3 SIRNA Assays ›
          Gene Symbol:
          Meg3
          Gene Name:
          maternally expressed 3
          Gene Aliases:
          2900016C05Rik, 3110050O07Rik, 6330408G06Rik, AI425946, AW108224, D12Bwg1266e, Gtl2, R74756, R75394
          Chromosome Location:
          Chr.12: 109540996 - 109571729 on Build GRCm38
          UniGene ID:
          Mm.289645
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n516113

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Meg3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003633.3
          2 1183
          NR_027651.2
          6 1171
          NR_027652.1
          7 1296
          GenBank mRNA AK018105.1 -
          2 1184
          AK147321.1 -
          4 2651
          BC024818.1 -
          4 543
          BC051667.1 -
          460
          BC094421.1 -
          6 1160
          Y13832.1 - LIT3453
          Gtl2
          7 1287
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CB194103.1 - 2 121 Chr. 12 - 109549289 - 109571721 [+] on Build GRCm38
          GenBankmRNA AW742476.1 - 3 286 Chr. 12 - 109544608 - 109558301 [+] on Build GRCm38
          GenBankmRNA CB183928.1 - 746 Chr. - - [-] on Build
          GenBankmRNA CF584712.1 - 500 Chr. - - [-] on Build
          GenBankmRNA CF746141.1 - 3 200 Chr. 12 - 109544694 - 109558925 [+] on Build GRCm38
          GenBankmRNA AA183389.1 - 2 141 Chr. 12 - 109549269 - 109558284 [+] on Build GRCm38
          GenBankmRNA BM053819.1 - 3 306 Chr. 12 - 109546356 - 109549803 [+] on Build GRCm38
          GenBankmRNA AV469420.1 - 346 Chr. - - [-] on Build
          GenBankmRNA BI730909.1 - 4 544 Chr. 12 - 109544446 - 109558310 [+] on Build GRCm38
          GenBankmRNA BF471019.1 - 3 404 Chr. 12 - 109544490 - 109558300 [+] on Build GRCm38
          GenBankmRNA BU058056.1 - 445 Chr. - - [-] on Build
          GenBankmRNA CK030896.1 - 4 401 Chr. 12 - 109544574 - 109571455 [+] on Build GRCm38
          GenBankmRNA BQ954402.1 - 1 59 Chr. 12 - 109549476 - 109559105 [+] on Build GRCm38
          GenBankmRNA BM729672.1 - 2 62 Chr. 12 - 109549348 - 109571555 [+] on Build GRCm38
          GenBankmRNA AA103529.1 - 1 26 Chr. 12 - 109549491 - 109571625 [+] on Build GRCm38
          GenBankmRNA AA004059.1 - 2 111 Chr. 12 - 109546470 - 109549810 [+] on Build GRCm38
          GenBankmRNA AV476864.1 - 2 124 Chr. 12 - 109549284 - 109571445 [+] on Build GRCm38
          GenBankmRNA CB202480.1 - 339 Chr. - - [-] on Build
          GenBankmRNA BF658505.1 - 2 155 Chr. 12 - 109549255 - 109571463 [+] on Build GRCm38
          GenBankmRNA BI441350.1 - 3 334 Chr. 12 - 109546328 - 109549691 [+] on Build GRCm38
          GenBankmRNA CB059150.1 - 2 190 Chr. 12 - 109549228 - 109571595 [+] on Build GRCm38
          GenBankmRNA AV584558.1 - 2 176 Chr. 12 - 109549234 - 109571423 [+] on Build GRCm38
          GenBankmRNA AV583825.1 - 4 287 Chr. 12 - 109544985 - 109558255 [+] on Build GRCm38
          GenBankmRNA CF584777.1 - 481 Chr. - - [-] on Build
          GenBankmRNA CF585122.1 - 3 362 Chr. 12 - 109546306 - 109571648 [+] on Build GRCm38
          GenBankmRNA CB590891.1 - 122 Chr. - - [-] on Build
          GenBankmRNA AA033444.1 - 2 140 Chr. 12 - 109549270 - 109558295 [+] on Build GRCm38
          GenBankmRNA CB058206.1 - 4 479 Chr. 12 - 109544498 - 109549697 [+] on Build GRCm38
          GenBankmRNA BI965904.1 - 3 306 Chr. 12 - 109546357 - 109549793 [+] on Build GRCm38
          GenBankmRNA AV480686.1 - 2 196 Chr. 12 - 109549224 - 109558320 [+] on Build GRCm38
          GenBankmRNA W64614.1 - 111 Chr. - - [-] on Build
          GenBankmRNA BM894630.1 - 2 122 Chr. 12 - 109549288 - 109571555 [+] on Build GRCm38
          GenBankmRNA BY718623.1 - 2 187 Chr. 12 - 109546394 - 109558814 [+] on Build GRCm38
          GenBankmRNA BM053951.1 - 3 306 Chr. 12 - 109546356 - 109549790 [+] on Build GRCm38
          GenBankmRNA AV502014.1 - 2 130 Chr. 12 - 109549280 - 109558307 [+] on Build GRCm38
          GenBankmRNA CF618073.1 - 2 277 Chr. 12 - 109549157 - 109571607 [+] on Build GRCm38
          GenBankmRNA BI965842.1 - 3 322 Chr. 12 - 109546340 - 109549809 [+] on Build GRCm38
          GenBankmRNA BE855234.1 - 2 221 Chr. 12 - 109549189 - 109558320 [+] on Build GRCm38

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: BC158675.

          Gene Ontology Categories:

          Function(s)

          in utero embryonic development
          liver development
          DNA methylation
          skeletal muscle tissue development
          determination of adult lifespan
          post-embryonic development
          multicellular organism growth
          lung alveolus development
          SMAD protein signal transduction
          genetic imprinting

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0