Search
Search

ATP11AUN| Gene Symbol: ATP11AUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207440.2 | NP_997323.1 | 3 | 1304 | ||
| GenBank mRNA | AK129953.1 | - | 3 | 1304 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000623100.2 | - | 1304 | Chr. - - [-] on Build | ||