Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUTM2B-AS1 SIRNA Assays ›
          Gene Symbol:
          NUTM2B-AS1
          Gene Name:
          NUTM2B antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 79691500 - 79826594 on Build GRCh38
          UniGene ID:
          Hs.440648
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n542260

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUTM2B-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_120611.1 4 603
          NR_120612.1 4 603
          NR_120613.1 4 603
          GenBank mRNA BQ425131.1 - 4 750
          CB163469.1 - 4 420
          CN279211.1 - 4 463
          CN279213.1 - 4 447
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AL520958.3 - 4 246 Chr. 10 - 79797377 - 79826237 [-] on Build GRCh38
          GenBankmRNA DA691562.1 - 4 486 Chr. 10 - 79804494 - 79826468 [-] on Build GRCh38
          GenBankmRNA BG111204.1 - 4 264 Chr. 10 - 79691671 - 79826255 [-] on Build GRCh38
          GenBankmRNA DA010830.1 - 5 537 Chr. 10 - 79806056 - 79826563 [-] on Build GRCh38
          GenBankmRNA BG166579.1 - 4 502 Chr. 10 - 79766137 - 79826484 [-] on Build GRCh38
          GenBankmRNA BG717953.1 - 4 538 Chr. 10 - 79691997 - 79826516 [-] on Build GRCh38
          GenBankmRNA CN274008.1 - 4 335 Chr. 10 - 79802853 - 79826326 [-] on Build GRCh38
          GenBankmRNA AV746236.1 - 2 107 Chr. 10 - 79766294 - 79808834 [-] on Build GRCh38
          GenBankmRNA AV745603.1 - 2 97 Chr. 10 - 79766118 - 79808828 [-] on Build GRCh38
          GenBankmRNA BG437437.1 - 571 Chr. - - [-] on Build
          GenBankmRNA BF437950.1 - 4 350 Chr. 10 - 79801984 - 79826341 [-] on Build GRCh38
          GenBankmRNA AW204120.1 - 4 400 Chr. 10 - 79804494 - 79826391 [-] on Build GRCh38
          GenBankmRNA BF223885.1 - 3 154 Chr. 10 - 79797859 - 79814658 [-] on Build GRCh38
          GenBankmRNA CN279215.1 - 4 487 Chr. 10 - 79804484 - 79826470 [-] on Build GRCh38
          GenBankmRNA AW612231.1 - 4 401 Chr. 10 - 79804492 - 79826392 [-] on Build GRCh38
          GenBankmRNA BM456581.1 - 748 Chr. - - [-] on Build
          GenBankmRNA BG256210.1 - 393 Chr. - - [-] on Build
          GenBankmRNA CN279212.1 - 4 441 Chr. 10 - 79804510 - 79826427 [-] on Build GRCh38
          GenBankmRNA BE742363.1 - 420 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0