Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other THAP9-AS1 SIRNA Assays ›
          Gene Symbol:
          THAP9-AS1 SEC31A
          Gene Name:
          THAP9 antisense RNA 1
          SEC31 homolog A, COPII coat complex component
          Gene Aliases:
          -
          ABP125, ABP130, HSPC275, HSPC334, SEC31L1
          Chromosome Location:
          Chr.4: 82893452 - 82900916 on Build GRCh38
          Chromosome Location:
          Chr.4: 82818509 - 82900571 on Build GRCh38
          UniGene ID:
          Hs.728916
          Hs.370024
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n542797

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: THAP9-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_034075.1 6 934
          NR_034076.1 5 909
          NR_034077.1 4 835
          GenBank mRNA AF278605.1 - 1 352
          AK128047.1 - 1 3077
          BC020725.1 - 5 540
          BC045722.1 - 4 1296
          DA361777.1 - 4 459
          U92014.1 - 3 134
          Gene Symbol: SEC31A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA DA142666.1 - 4 481
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA833028.1 - 5 561 Chr. 4 - 82894668 - 82900566 [-] on Build GRCh38
          GenBankmRNA BU178413.1 - 4 395 Chr. 4 - 82894385 - 82900476 [-] on Build GRCh38
          GenBankmRNA BX339948.2 - 5 542 Chr. 4 - 82894370 - 82900549 [-] on Build GRCh38
          GenBankmRNA BE880640.1 - 4 423 Chr. 4 - 82894534 - 82900504 [-] on Build GRCh38
          GenBankmRNA T98067.1 - 134 Chr. - - [-] on Build
          GenBankmRNA CB127731.1 - 5 500 Chr. 4 - 82894723 - 82900507 [-] on Build GRCh38
          GenBankmRNA DA925660.1 - 5 500 Chr. 4 - 82894740 - 82900506 [-] on Build GRCh38
          GenBankmRNA DB476991.1 - 4 486 Chr. 4 - 82894795 - 82900566 [-] on Build GRCh38
          GenBankmRNA BI762414.1 - 5 529 Chr. 4 - 82894631 - 82900536 [-] on Build GRCh38
          GenBankmRNA BU506989.1 - 5 467 Chr. 4 - 82894394 - 82900474 [-] on Build GRCh38
          GenBankmRNA DA568200.1 - 5 540 Chr. 4 - 82894620 - 82900547 [-] on Build GRCh38
          GenBankmRNA BU175480.1 - 4 276 Chr. 4 - 82894208 - 82900173 [-] on Build GRCh38
          GenBankmRNA DA055649.1 - 4 488 Chr. 4 - 82894754 - 82900566 [-] on Build GRCh38
          GenBankmRNA DA912133.1 - 4 427 Chr. 4 - 82894681 - 82900506 [-] on Build GRCh38
          GenBankmRNA DA615125.1 - 5 526 Chr. 4 - 82894781 - 82900536 [-] on Build GRCh38
          GenBankmRNA BF886678.1 - 4 288 Chr. 4 - 82894665 - 82900159 [-] on Build GRCh38
          GenBankmRNA CN429666.1 - 5 482 Chr. 4 - 82894685 - 82900483 [-] on Build GRCh38
          GenBankmRNA BM854572.1 - 3 122 Chr. 4 - 82894455 - 82898005 [-] on Build GRCh38
          GenBankmRNA CN429665.1 - 4 380 Chr. 4 - 82894541 - 82900461 [-] on Build GRCh38
          GenBankmRNA AL603040.1 - 5 347 Chr. 4 - 82894613 - 82900354 [-] on Build GRCh38
          GenBankmRNA CN429663.1 - 5 415 Chr. 4 - 82894757 - 82900413 [-] on Build GRCh38
          GenBankmRNA BM478668.1 - 481 Chr. - - [-] on Build
          GenBankmRNA DA082618.1 - 4 487 Chr. 4 - 82894739 - 82900566 [-] on Build GRCh38
          GenBankmRNA DA356680.1 - 4 431 Chr. 4 - 82894685 - 82900509 [-] on Build GRCh38
          GenBankmRNA BM472827.1 - 488 Chr. - - [-] on Build
          GenBankmRNA R11369.1 - 3 135 Chr. 4 - 82894681 - 82898017 [-] on Build GRCh38
          GenBankmRNA AL514590.3 - 5 451 Chr. 4 - 82894205 - 82900459 [-] on Build GRCh38
          GenBankmRNA DB030130.1 - 5 490 Chr. 4 - 82894751 - 82900496 [-] on Build GRCh38
          GenBankmRNA DA958580.1 - 5 502 Chr. 4 - 82894765 - 82900506 [-] on Build GRCh38
          GenBankmRNA DB257486.1 - 5 529 Chr. 4 - 82894782 - 82900533 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0