Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LINC-PINT SIRNA Assays ›
          Gene Symbol:
          LINC-PINT
          Gene Name:
          long intergenic non-protein coding RNA, p53 induced transcript
          Gene Aliases:
          LincRNA-Pint, MKLN1-AS1, PINT
          Chromosome Location:
          Chr.7: 130877562 - 131109916 on Build GRCh38
          UniGene ID:
          Hs.150556
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n542983

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LINC-PINT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015431.2 4 1421
          NR_024153.2 3 387
          NR_109850.1 5 1545
          NR_109851.1 4 1421
          NR_109852.1 4 1421
          NR_109853.1 4 1421
          NR_109854.1 4 1421
          NR_109855.1 3 1290
          GenBank mRNA AK125651.1 - 5 1523
          AL713664.1 - 3 238
          AV727262.1 - 3 368
          BC039420.1 - 3 387
          BC047367.1 - 3 567
          BC130416.1 - 3 1089
          BG706681.1 - 3 537
          BI833488.1 - 3 417
          BI914823.1 - 3 370
          CD673688.1 - 2 169
          DA182364.1 - 3 230
          DA321983.1 - 2 370
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI667964.1 - 3 541 Chr. 7 - 130945736 - 131108097 [-] on Build GRCh38
          GenBankmRNA BM142071.1 - 3 543 Chr. 7 - 130984074 - 131108105 [-] on Build GRCh38
          GenBankmRNA AV735232.1 - 3 271 Chr. 7 - 130944721 - 131107834 [-] on Build GRCh38
          GenBankmRNA BI755237.1 - 3 497 Chr. 7 - 130945587 - 131108059 [-] on Build GRCh38
          GenBankmRNA DA013764.1 - 3 573 Chr. 7 - 130984070 - 131108135 [-] on Build GRCh38
          GenBankmRNA DA164941.1 - 3 535 Chr. 7 - 130984049 - 131108097 [-] on Build GRCh38
          GenBankmRNA BM548007.1 - 3 491 Chr. 7 - 130945395 - 131108034 [-] on Build GRCh38
          GenBankmRNA BM808513.1 - 3 570 Chr. 7 - 130944672 - 131108110 [-] on Build GRCh38
          GenBankmRNA BI597250.1 - 3 392 Chr. 7 - 130944637 - 131107030 [-] on Build GRCh38
          GenBankmRNA DB265374.1 - 3 434 Chr. 7 - 130945715 - 131107983 [-] on Build GRCh38
          GenBankmRNA BM469529.1 - 480 Chr. - - [-] on Build
          GenBankmRNA CK822014.1 - 3 469 Chr. 7 - 130984049 - 131108031 [-] on Build GRCh38
          GenBankmRNA DA014861.1 - 3 535 Chr. 7 - 130984055 - 131108097 [-] on Build GRCh38
          GenBankmRNA BI461845.1 - 3 585 Chr. 7 - 130945641 - 131108145 [-] on Build GRCh38
          GenBankmRNA BI764213.1 - 3 521 Chr. 7 - 130945574 - 131108082 [-] on Build GRCh38
          GenBankmRNA DB113889.1 - 3 494 Chr. 7 - 130945785 - 131108056 [-] on Build GRCh38
          GenBankmRNA BG719956.1 - 3 585 Chr. 7 - 130945718 - 131108145 [-] on Build GRCh38
          GenBankmRNA BI668071.1 - 3 566 Chr. 7 - 130945715 - 131108109 [-] on Build GRCh38
          GenBankmRNA BI768447.1 - 3 262 Chr. 7 - 130872235 - 131052536 [-] on Build GRCh38
          GenBankmRNA AL542592.3 - 3 563 Chr. 7 - 130945464 - 131108123 [-] on Build GRCh38
          GenBankmRNA CB988481.1 - 3 590 Chr. 7 - 130967430 - 131108133 [-] on Build GRCh38
          GenBankmRNA BI755625.1 - 3 265 Chr. 7 - 130944567 - 131107827 [-] on Build GRCh38
          GenBankmRNA BQ220972.1 - 566 Chr. - - [-] on Build
          GenBankmRNA DB497590.1 - 3 385 Chr. 7 - 130945748 - 131107028 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0